Learn More
Medchemexpress LLC Duloxetine metabolite Para-Naphthol Duloxetine | 949095-98-1 | >98% | 297.41 | 10 MG
SDP
Catalog No. 5000365089
Encompass_Preferred
Para-Naphthol Duloxetine is a metabolite of Duloxetine, which is a serotonin-norepinephrine reuptake inhibitor (SNRI).
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C, 1 month at -20°C
  • Targeted as a drug metabolite
  • Pathway is metabolic enzyme/protease
Catalog No. 50-003-65089 Supplier Medchemexpress LLC Supplier No. 10MGHYG0005
Para-Naphthol Duloxetine is a metabolite of Duloxetine, which is a serotonin-norepinephrine reuptake inhibitor (SNRI).
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C, 1 month at -20°C
  • Targeted as a drug metabolite
  • Pathway is metabolic enzyme/protease