Learn More
Fine Science Tools Dumont #L5 - Sliding Lock Forceps
SDP
Catalog No. NC9217669
Encompass
Style: #L5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC9217669 Supplier Fine Science Tools Supplier No. 1125327
Style: #L5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm