Learn More
Electron Microscopy Sciences Spinbar Circulus™ Dumbbell Stiriing Bar 32mm Blue
SDP
Catalog No. 501901851
Encompass_Preferred
These magnet stirring bars provide strong turbulence at relatively low speeds, offering reduced surface contact and excellent centering characteristics, particularly in vessels with convex bottoms. Ideal for 'problem' liquids, highly viscous mixtures and metal particles in fluid. All sizes have solid PTFE end disks 20 mm (.79”) in diameter. Bar diameter is 8 mm (.315”).
Color: Blue
Catalog No. 50-190-1851 Supplier Electron Microscopy Sciences Supplier No. 64511B
These magnet stirring bars provide strong turbulence at relatively low speeds, offering reduced surface contact and excellent centering characteristics, particularly in vessels with convex bottoms. Ideal for 'problem' liquids, highly viscous mixtures and metal particles in fluid. All sizes have solid PTFE end disks 20 mm (.79”) in diameter. Bar diameter is 8 mm (.315”).
Color: Blue