Learn More
Electron Microscopy Sciences Spinbar Circulus™ Dumbbell Stiriing Bar 52mm Blue
SDP
Catalog No. 501901855
Encompass_Preferred
These magnet stirring bars provide strong turbulence at relatively low speeds, offering reduced surface contact and excellent centering characteristics, particularly in vessels with convex bottoms. Ideal for 'problem' liquids, highly viscous mixtures and metal particles in fluid. All sizes have solid PTFE end disks 20 mm (.79”) in diameter. Bar diameter is 8 mm (.315”).
Color: Blue
Catalog No. 50-190-1855 Supplier Electron Microscopy Sciences Supplier No. 64512B
These magnet stirring bars provide strong turbulence at relatively low speeds, offering reduced surface contact and excellent centering characteristics, particularly in vessels with convex bottoms. Ideal for 'problem' liquids, highly viscous mixtures and metal particles in fluid. All sizes have solid PTFE end disks 20 mm (.79”) in diameter. Bar diameter is 8 mm (.315”).
Color: Blue