Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RWD LIFE SCIENCE INC Dummy Cannula (Plastic)-Single/OD 0.30mm/Mates with M3.5
SDP

Catalog No. NC3393384
Encompass

Dummy Cannula (Plastic)-Single/OD 0.30mm/Mates with M3.5

Catalog No. NC3393384 Supplier RWD LIFE SCIENCE INC Supplier No. 621028MM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dummy Cannula (Plastic)-Single/OD 0.30mm/Mates with M3.5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.