Learn More
World Precision Instrument Dummy Load Resistor Kit
SDP
Catalog No. 502538864
Encompass
A365 can be used as a precise voltage source by placing a precision “dummy” resistor load across the output terminals. Precision resistors (such as the 100 Ω, 1000 Ω and 10,000 Ω resistors in WPI’s optional Dummy Load Resistor Kit #DRL) when placed across the output terminals convert the precision current levels to accurate voltage levels.
For example, 1 milliampere flowing through 1000 Ohms will produce 1 volt across the output terminal pair. Thus, models A365D and A365R can be used as low noise sources of accurate voltage pulses or DC. To maintain less than 1% error, avoid shunting the output dummy resistor with a load resistance less than 100 times the dummy resistance value.
Catalog No. 50-253-8864 Supplier World Precision Instrument Supplier No. DRL
A365 can be used as a precise voltage source by placing a precision “dummy” resistor load across the output terminals. Precision resistors (such as the 100 Ω, 1000 Ω and 10,000 Ω resistors in WPI’s optional Dummy Load Resistor Kit #DRL) when placed across the output terminals convert the precision current levels to accurate voltage levels.
For example, 1 milliampere flowing through 1000 Ohms will produce 1 volt across the output terminal pair. Thus, models A365D and A365R can be used as low noise sources of accurate voltage pulses or DC. To maintain less than 1% error, avoid shunting the output dummy resistor with a load resistance less than 100 times the dummy resistance value.