Learn More
Roboz Surgical Dumont Tweezers; Pattern #5; .08 X .04 mm Tip Size; 110 mm Length; Carbon Steel
SDP
Catalog No. NC0410614
Encompass
Dumont Tweezers; Pattern #5; .08 X .04mm Tip Size; 110mm Length; Material: Carbon Steel Carbon steel is an extremely hard alloy (> 59 HRC.) However, the carbon steel material has no corrosion protection in its composition. It is normal for this mate
Catalog No. NC0410614 Supplier Roboz Surgical Supplier No. RS5035
Dumont Tweezers; Pattern #5; .08 X .04mm Tip Size; 110mm Length; Material: Carbon Steel Carbon steel is an extremely hard alloy (> 59 HRC.) However, the carbon steel material has no corrosion protection in its composition. It is normal for this mate