Learn More
Fine Science Tools Dumont #5 - Mirror Finish Forceps, 11 cm
SDP
Catalog No. NC1856015
Encompass
These forceps are highly polished to prevent embryos, nerves, or other adhesive tissues from sticking to the tips.
Catalog No. NC1856015 Supplier Fine Science Tools Supplier No. 1125223
These forceps are highly polished to prevent embryos, nerves, or other adhesive tissues from sticking to the tips.