Learn More
Fine Science Tools DUMONT AA INOX FRCP
SDP
Catalog No. NC1470608
Encompass
Style: AA/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.4 x 0.2mm/ Alloy / Material: Inox/ Length: 12.5cm
Catalog No. NC1470608 Supplier Fine Science Tools Supplier No. 1121020
Style: AA/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.4 x 0.2mm/ Alloy / Material: Inox/ Length: 12.5cm