Learn More
Fine Science Tools Dumont SS Forceps, 13.5cm
SDP
Catalog No. NC9504087
Encompass
Style: SS/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.2 x 0.12mm/ Alloy / Material: Inox/ Length: 13.5cm. The tips are smooth on the inside for a gentle grip. Applying full strength will not separate tips and a strong hold on the smallest specimen is ensured.
Catalog No. NC9504087 Supplier Fine Science Tools Supplier No. 1120323
Style: SS/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.2 x 0.12mm/ Alloy / Material: Inox/ Length: 13.5cm. The tips are smooth on the inside for a gentle grip. Applying full strength will not separate tips and a strong hold on the smallest specimen is ensured.