Learn More
Braintree Scientific Dumont Forceps, 45 degree, tip .1x.06, 109mm Dumoxel
SDP
Catalog No. 501954832
Encompass
Dumont Tweezers; Pattern #5, 45 Degree Angle; .10 X .06mm Tip Size; 109mm Length; Material: Dumoxel **Sold for research purposes only and not for human use**
Catalog No. 50-195-4832 Supplier Braintree Scientific Supplier No. FC5058
Dumont Tweezers; Pattern #5, 45 Degree Angle; .10 X .06mm Tip Size; 109mm Length; Material: Dumoxel **Sold for research purposes only and not for human use**