Learn More
Fine Science Tools Dumont #3 Forceps, 12cm
SDP
Catalog No. NC9341131
Encompass
Style: #3/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumoxel/ Length: 12cm
Catalog No. NC9341131 Supplier Fine Science Tools Supplier No. 1123130
Style: #3/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumoxel/ Length: 12cm