Learn More
Fine Science Tools DUMONT FORCEPS NO5-45 11CM
SDP
Catalog No. NC9341129
Encompass
Style: #5/45/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumoxel/ Length: 11cm
Catalog No. NC9341129 Supplier Fine Science Tools Supplier No. 1125135
Style: #5/45/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumoxel/ Length: 11cm