Learn More
Fine Science Tools Dumont Forceps - Micro-Blunted Tips, Curved, 0.17 x 0.1 mm, 11.5 cm
SDP
Catalog No. NC9402871
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Inox/ Length: 11.5cm
Catalog No. NC9402871 Supplier Fine Science Tools Supplier No. 1127320
Style: #7/ Tip Shape: Curved/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Inox/ Length: 11.5cm