Learn More
Braintree Scientific Dumont Inox Forceps,109mm Inox, 45 degree, .05/.01mm tip
SDP
Catalog No. 501954829
Encompass
Dumont Tweezers; Pattern #5, 45 Degree Angle; .05 X .01mm Tip Size; 109mm Length; Material: Inox; Biologie Tips **Sold for research purposes only and not for human use**
Catalog No. 50-195-4829 Supplier Braintree Scientific Supplier No. FC5005
 Restricted Item
Add to Cart
Add to Cart
Dumont Tweezers; Pattern #5, 45 Degree Angle; .05 X .01mm Tip Size; 109mm Length; Material: Inox; Biologie Tips **Sold for research purposes only and not for human use**