Learn More
Fine Science Tools Dumont Forceps - Micro-Blunted Tips, Angled 90°, 0.1 x 0.06 mm, 11 cm
SDP
Catalog No. NC9352180
Encompass
Style: #5/90/ Tip Shape: Angled 90 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC9352180 Supplier Fine Science Tools Supplier No. 1125329
Style: #5/90/ Tip Shape: Angled 90 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm