Learn More
Fine Science Tools Dumont Mini Forceps - Style 3
SDP
Catalog No. NC2002347
Encompass
Dumont Mini Forceps - Style 3 | Style: Mini #3, Tip Shape: Straight, Tips: StandardTip, Dimensions:0.12 x 0.07mm, Alloy / Material:Inox, Length: 7cm
Catalog No. NC2002347 Supplier Fine Science Tools Supplier No. 1120010
Dumont Mini Forceps - Style 3 | Style: Mini #3, Tip Shape: Straight, Tips: StandardTip, Dimensions:0.12 x 0.07mm, Alloy / Material:Inox, Length: 7cm