Learn More
Roboz Surgical Dumont Tweezers
SDP
Catalog No. NC0905062
Encompass
Dumont Tweezers; Pattern #N5; Cross Action; Thin Tips; .05 X .01mm Tip Size; 108mm Length; Material: Inox; Biologie Tips.
Catalog No. NC0905062 Supplier Roboz Surgical Supplier No. RS5020
Dumont Tweezers; Pattern #N5; Cross Action; Thin Tips; .05 X .01mm Tip Size; 108mm Length; Material: Inox; Biologie Tips.