Learn More
Fine Science Tools Dumont #3 Forceps, Straight, 0.17 x 0.1 mm, 12 cm
SDP
Catalog No. NC9858013
Encompass
Style: #3/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumostar/ Length: 12cm
Catalog No. NC9858013 Supplier Fine Science Tools Supplier No. 1129300
Style: #3/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.17 x 0.1mm/ Alloy / Material: Dumostar/ Length: 12cm