Learn More
Fine Science Tools Dumont Forceps - Micro-Blunted Tips
SDP
Catalog No. NC9779941
Encompass
Style: #5/45/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm. These forceps have atraumatic or gently blunted tips to minimize the chance of puncturing tissue.
Catalog No. NC9779941 Supplier Fine Science Tools Supplier No. 1125325
Style: #5/45/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm. These forceps have atraumatic or gently blunted tips to minimize the chance of puncturing tissue.