Learn More
Andwin Scientific DUMONT NO5 DUMOXEL
SDP
Catalog No. NC9533247
Encompass_Preferred
The Dumont tweezer Style #5 features super thin polished tips. This product has an overall length of 110 millimeters. It is sold individually.
Catalog No. NC9533247 Supplier Andwin Scientific Supplier No. 72700D
The Dumont tweezer Style #5 features super thin polished tips. This product has an overall length of 110 millimeters. It is sold individually.