Learn More
Fine Science Tools Dumont Forceps - Micro-Blunted Tips, Straight, 0.1 x 0.06 mm, 11 cm
SDP
Supplier:  Fine Science Tools 1125320
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC9521913