Learn More
Fine Science Tools DUMONT NO5 STUDENT QUALITY
SDP
Catalog No. NC9277114
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm/ Feature: Student Quality
Catalog No. NC9277114 Supplier Fine Science Tools Supplier No. 9115020
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Inox/ Length: 11cm/ Feature: Student Quality