Learn More
Fine Science Tools Dumont #55 Forceps, Straight, Dumostar, 0.05 x 0.02 mm, 11 cm
SDP
Catalog No. NC9504088
Encompass
Style: #55/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.02mm/ Alloy / Material: Dumostar/ Length: 11cm
Catalog No. NC9504088 Supplier Fine Science Tools Supplier No. 1129551
Style: #55/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.02mm/ Alloy / Material: Dumostar/ Length: 11cm