Learn More
Fine Science Tools DUMONT NO5ST INOX
SDP
Catalog No. NC9473431
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.01mm/ Alloy / Material: Inox/ Length: 11cm
Catalog No. NC9473431 Supplier Fine Science Tools Supplier No. 1125420
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.01mm/ Alloy / Material: Inox/ Length: 11cm