Learn More
Roboz Surgical DUMONT NO7 TITANIUM .07X.03MM
SDP
Catalog No. NC9593817
Encompass
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Titanium; Biologie Tips
Catalog No. NC9593817 Supplier Roboz Surgical Supplier No. RS5039
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Titanium; Biologie Tips