Learn More
Fine Science Tools Dumont SS Forceps, Angled 45°, Inox, 0.2 x 0.12 mm, 13.5 cm
SDP
Catalog No. NC9442576
Encompass
Style: SS/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.2 x 0.12mm/ Alloy / Material: Inox/ Length: 13.5cm
Catalog No. NC9442576 Supplier Fine Science Tools Supplier No. 1120325
Style: SS/ Tip Shape: Angled 45 deg./ Tips: Standard/ Tip Dimensions: 0.2 x 0.12mm/ Alloy / Material: Inox/ Length: 13.5cm