Learn More
Mitegen Style 5 Tweezers by Dumont, Super Thin Tips (0.01x0.05mm), 110mm Length, Polished Dumoxel Alloy, Biological Grade, Precision Forceps for Micro-Manipulation
SDP
Catalog No. NC2675831
Encompass
The Dumont Style 5 Tweezers with Super Thin Tips are precision instruments for delicate micro-manipulation in scientific and industrial applications. Made from polished, high-quality Dumoxel alloy, these biological grade tweezers resist acids and corrosion, ensuring lasting performance and easy cleaning in demanding lab environments. Their extremely fine tips (0.01 x 0.05 mm) provide unparalleled precision for handling minute samples, fragile grids, or delicate components without damage. At 110 mm long, they offer excellent balance and control, reducing hand fatigue during intricate use. Ideal for electron microscopy (EM), crystallography, cell culture, and micro-assembly, these tweezers are an indispensable tool where accuracy, sample integrity, and durability are paramount.
Catalog No. NC2675831 Supplier Mitegen Supplier No. MCEM72700D
The Dumont Style 5 Tweezers with Super Thin Tips are precision instruments for delicate micro-manipulation in scientific and industrial applications. Made from polished, high-quality Dumoxel alloy, these biological grade tweezers resist acids and corrosion, ensuring lasting performance and easy cleaning in demanding lab environments. Their extremely fine tips (0.01 x 0.05 mm) provide unparalleled precision for handling minute samples, fragile grids, or delicate components without damage. At 110 mm long, they offer excellent balance and control, reducing hand fatigue during intricate use. Ideal for electron microscopy (EM), crystallography, cell culture, and micro-assembly, these tweezers are an indispensable tool where accuracy, sample integrity, and durability are paramount.