Learn More
Electron Microscopy Sciences Dumont Tweezers Positive Action Style 5/15, High Precision, Dumoxel, Polished Finish, OAL
SDP
Catalog No. 5024112
Encompass_Preferred
Style 5/15 tweezers have thin tips that are bent 15 degrees, for hairsprings.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and offers the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 degree C (DIN 50 914) and is suitable for autoclave sterilization at 270 degree C.
Grade: High Precision
Material: Dumoxel
Finish: Polished
Catalog No. 50-241-12 Supplier Electron Microscopy Sciences Supplier No. 72708DZ
Style 5/15 tweezers have thin tips that are bent 15 degrees, for hairsprings.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and offers the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 degree C (DIN 50 914) and is suitable for autoclave sterilization at 270 degree C.
Grade: High Precision
Material: Dumoxel
Finish: Polished