Learn More
Fine Science Tools Dumont #7 - Ceramic Coated Forceps, Curved, 0.07 x 0.04 mm, 11 cm
SDP
Catalog No. NC0191963
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Biologie/ Tip Dimensions: 0.07 x 0.04mm/ Alloy / Material: Ceramic Coated Inox/ Length: 11cm
Catalog No. NC0191963 Supplier Fine Science Tools Supplier No. 1127250
Style: #7/ Tip Shape: Curved/ Tips: Biologie/ Tip Dimensions: 0.07 x 0.04mm/ Alloy / Material: Ceramic Coated Inox/ Length: 11cm