Learn More
Fine Science Tools Dumont #4 Forceps, Straight, 0.13 x 0.08 mm, 11 cm
SDP
Catalog No. NC9959687
Encompass
Style: #4/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.13 x 0.08mm/ Alloy / Material: Dumostar/ Length: 11cm
Catalog No. NC9959687 Supplier Fine Science Tools Supplier No. 1129400
Style: #4/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.13 x 0.08mm/ Alloy / Material: Dumostar/ Length: 11cm