Learn More
Fine Science Tools Dumont #5CO Forceps, Straight, 0.05 x 0.01 mm, 11 cm
SDP
Catalog No. NC9346566
Encompass
Style: #5CO/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.01mm/ Alloy / Material: Dumostar/ Length: 11cm
Catalog No. NC9346566 Supplier Fine Science Tools Supplier No. 1129520
Style: #5CO/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.01mm/ Alloy / Material: Dumostar/ Length: 11cm