Learn More
Fine Science Tools Dumont Mini Forceps - Style 5, Straight, 0.09 x 0.05 mm, 8.2 cm
SDP
Catalog No. NC9004323
Encompass
Style: Mini #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.09 x 0.05mm/ Alloy / Material: Inox/ Length: 8.2cm
Catalog No. NC9004323 Supplier Fine Science Tools Supplier No. 1120014
Style: Mini #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.09 x 0.05mm/ Alloy / Material: Inox/ Length: 8.2cm