Learn More
Fine Science Tools Dumont #4 Forceps, Straight, 0.06 x 0.04 mm, 11 cm
SDP
Catalog No. NC9750192
Encompass
Style: #4/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.06 x 0.04mm/ Alloy / Material: Dumoxel/ Length: 11cm
Catalog No. NC9750192 Supplier Fine Science Tools Supplier No. 1124240
Style: #4/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.06 x 0.04mm/ Alloy / Material: Dumoxel/ Length: 11cm