Learn More
MSC BETCO Dumpster Treatment, Odor Eliminator, Probiotic, Mango Scent, 1 Gallon
Catalog No. 502839384
Encompass
Betco Dumpster Treatment is a powerful solution designed for effective cleaning and odor elimination in dumpsters and other trash receptacles. It provides excellent clinging foam for extended contact and maximum cleaning and elimination of malodors. Regular use eliminates malodors. It can also be used on trash chutes and loading docks.
  • Rich clinging foam for extended cleaning and odor control.
  • Triple odor control including fragrance, malodor counteractant, and probiotic bacteria.
  • Bacteria count of 325 billion CFU per gallon.
  • Pleasant mango fragrance.
  • Yellow in color with a pH range of 6.5-7.5.
  • Excellent grease emulsification.
  • High foaming action.
Catalog No. 50-283-9384 Supplier MSC Supplier No. 73004533
Betco Dumpster Treatment is a powerful solution designed for effective cleaning and odor elimination in dumpsters and other trash receptacles. It provides excellent clinging foam for extended contact and maximum cleaning and elimination of malodors. Regular use eliminates malodors. It can also be used on trash chutes and loading docks.
  • Rich clinging foam for extended cleaning and odor control.
  • Triple odor control including fragrance, malodor counteractant, and probiotic bacteria.
  • Bacteria count of 325 billion CFU per gallon.
  • Pleasant mango fragrance.
  • Yellow in color with a pH range of 6.5-7.5.
  • Excellent grease emulsification.
  • High foaming action.