Learn More
MSC Dunlop Chest Wader, PVC, Steel Toe, Size 13, 1 ea
Catalog No. 5000793090
Encompass
This Hip & Chest Wader features a high-leg boot design, constructed from durable PVC in black, and is a men's size 13. It includes robust protection with a steel toe and steel lining, designed for demanding environments.
  • High-leg boot style
  • Chest wader type
  • Steel toe for impact protection
  • Steel lining for durability
  • Non-slip PVC outsole
  • Waterproof and cold protection features
  • Pull-on closure
  • Complies with ASTM F2413-11 M I/75 C/75 EH standards
  • Medium shoe width
Catalog No. 50-007-93090 Supplier MSC Supplier No. 60932944
This Hip & Chest Wader features a high-leg boot design, constructed from durable PVC in black, and is a men's size 13. It includes robust protection with a steel toe and steel lining, designed for demanding environments.
  • High-leg boot style
  • Chest wader type
  • Steel toe for impact protection
  • Steel lining for durability
  • Non-slip PVC outsole
  • Waterproof and cold protection features
  • Pull-on closure
  • Complies with ASTM F2413-11 M I/75 C/75 EH standards
  • Medium shoe width