Learn More
MSC Dunlop Hip Wader, PVC, Steel Toe, Size 6, 1 ea
Catalog No. 5000794389
Encompass
This hip and chest wader features a high-leg boot design, constructed from durable black PVC. It is engineered for demanding environments, offering reliable protection and comfort for male users.
  • Equipped with a steel toe for enhanced safety.
  • Non-slip sole provides secure footing.
  • Waterproof construction offers superior resistance in wet conditions.
  • Provides cold protection.
  • Designed with a convenient pull-on closure.
  • Meets ASTM F2413-11 M I/75 C/75 EH standards.
Catalog No. 50-007-94389 Supplier MSC Supplier No. 60932951
This hip and chest wader features a high-leg boot design, constructed from durable black PVC. It is engineered for demanding environments, offering reliable protection and comfort for male users.
  • Equipped with a steel toe for enhanced safety.
  • Non-slip sole provides secure footing.
  • Waterproof construction offers superior resistance in wet conditions.
  • Provides cold protection.
  • Designed with a convenient pull-on closure.
  • Meets ASTM F2413-11 M I/75 C/75 EH standards.