Learn More
MSC Dunlop Hip Wader, Steel Toe, Men's Size 9, 1 ea
Catalog No. 5000782480
Encompass
Dunlop Protective Footwear hip waders are designed for demanding environments, offering reliable protection and comfort. These high-leg boots feature a steel toe and are made from durable PVC, making them suitable for various applications requiring foot protection and water resistance.
  • High-leg boot style
  • Steel toe type
  • Men's size 9
  • Black color
  • Wader height of 35
  • PVC material construction
  • Non-slip sole type
  • PVC outsole material
  • Meets ASTM F2413-11 M I/75 C/75 EH standards
  • Medium shoe width
  • Waterproof and cold protection features
  • Pull-on closure type
Catalog No. 50-007-82480 Supplier MSC Supplier No. 60932985
Dunlop Protective Footwear hip waders are designed for demanding environments, offering reliable protection and comfort. These high-leg boots feature a steel toe and are made from durable PVC, making them suitable for various applications requiring foot protection and water resistance.
  • High-leg boot style
  • Steel toe type
  • Men's size 9
  • Black color
  • Wader height of 35
  • PVC material construction
  • Non-slip sole type
  • PVC outsole material
  • Meets ASTM F2413-11 M I/75 C/75 EH standards
  • Medium shoe width
  • Waterproof and cold protection features
  • Pull-on closure type