Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Dunlop Protective Footwear Rain Jacket, Medium, ASTM D6413, 1 ea

Catalog No. 5000787670
Encompass

This rain jacket is designed for protection in non-hazardous environments, featuring a durable polyester construction in a green finish. It meets ASTM D6413 standards, ensuring reliability and performance for various outdoor tasks.

  • Sleeve length of 34.00 in
  • Attached hood
  • Chest size of 50 in
  • Top-stitched, double-stitched, and taped seams
  • Mildew-resistant
  • Open wrist style
  • Safetyflex protection series for non-hazardous protection
  • Material weight of 13 oz.
  • Material thickness of 17.0 mm
  • Snaps and zipper closure

Catalog No. 50-007-87670 Supplier MSC Supplier No. 70137278
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rain jacket is designed for protection in non-hazardous environments, featuring a durable polyester construction in a green finish. It meets ASTM D6413 standards, ensuring reliability and performance for various outdoor tasks.

  • Sleeve length of 34.00 in
  • Attached hood
  • Chest size of 50 in
  • Top-stitched, double-stitched, and taped seams
  • Mildew-resistant
  • Open wrist style
  • Safetyflex protection series for non-hazardous protection
  • Material weight of 13 oz.
  • Material thickness of 17.0 mm
  • Snaps and zipper closure

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.