Learn More
Grainger Dunnage Rack 37 3/4 in L 24 in W
Catalog No. 502752867
Encompass_Preferred
The Grainger 14G286 is a 14 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The hammer has a fiberglass handle that provides durability. This general purpose hammer allows the user to deliver forceful blows with minimal recoil. It is well-suited for assembling equipment or driving machine parts without marring the surface. The compact size makes it easy to handle in tight spaces. With proper care, this hammer will provide reliable striking performance.
Catalog No. 50-275-2867 Supplier Grainger Supplier No. 14G286
The Grainger 14G286 is a 14 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The hammer has a fiberglass handle that provides durability. This general purpose hammer allows the user to deliver forceful blows with minimal recoil. It is well-suited for assembling equipment or driving machine parts without marring the surface. The compact size makes it easy to handle in tight spaces. With proper care, this hammer will provide reliable striking performance.