Learn More
Diversified Spaces™ Dunnage Racks
SCI_ED
Catalog No. S37870 Shop All Diversified Spaces Products
Change view
Click to view available options
Material:
Aluminum Tubing
Catalog No. Material
S37870 Aluminum Tubing
1 options
Catalog No. S37870 Supplier Diversified Spaces™ Supplier No. 250489
To store heavy and bulky loads, keeping them off the ground
  • Made from 1.5 in. corrosion-resistant square aluminum tubing
  • Square tube frames provide extra strength
  • 36 in. W model holds up to 3000 lb.
Specifications
Material Aluminum Tubing
For Use With (Application) store heavy and bulky loads
Size D x W x H: 20 x 36 x 12 in.