Learn More
World Precision Instrument Duo 773 Electrometer
SDP
Catalog No. 502538506
Encompass
2-Channel intracellular amplifier for dual and differential studies
The Duo773 has two channes for differential or intracellular ISE with an integrated DC current generater and external control input. It has an integrated low pass filter. The bridge balance circuit nullifies the electrode voltage drop. And, it has an integrated "tickler" circuit with patent-pending electrode flutter technology. It has integrated test ports for each channel and dual capacitance compensations with output offse controls. The Duo773 comes complete with two probe headstages, normally one high impedance ona one low impedance.
Catalog No. 50-253-8506 Supplier World Precision Instrument Supplier No. SYS773
Add to Cart
Add to Cart
Request a Quote
2-Channel intracellular amplifier for dual and differential studies
The Duo773 has two channes for differential or intracellular ISE with an integrated DC current generater and external control input. It has an integrated low pass filter. The bridge balance circuit nullifies the electrode voltage drop. And, it has an integrated "tickler" circuit with patent-pending electrode flutter technology. It has integrated test ports for each channel and dual capacitance compensations with output offse controls. The Duo773 comes complete with two probe headstages, normally one high impedance ona one low impedance.