Learn More
Thermo Scientific DUO CERAMIC CONE
Supplier:  Thermo Scientific 842312051261
Encompass
Duo Ceramic Cone; Replaceable ceramic cone forming the purgable interface between the; plasma and optical system.; For use with iCAP 6000 Series Duo instruments only.
Catalog No. NC9063742