Learn More
Med Vet International My First Lab Mega DUO-SCOPE
Supplier:  Med Vet International MFL26
Encompass_Preferred
Mega Duo Scope (Model MFL-26) expands our most popular duo scope in the My First Lab series! Two light sources allow this microscope to magnify both slides and solid objects! View slides, coins, plants, stamps, insects, jewelry and more!You would need two instruments to view what this one does a basic biological scope to look at specimens on a slide (light shines up from under the slide and through the tissue) and a traditional stereo or dissecting microscope to view solid objects at lower power (light shines down onto the specimen to be observed). This new Duo Scope with dual lighting allows for both types of applications! Cordless battery power makes it possible for field study. Priced to fit any budget with the same fine quality as our other scopes.
Catalog No. 50-304-6846