Learn More
Azenta US Inc DUO SINGLE TUBE SCANNER 2D/1D
Catalog No. NC3585175
Encompass
Duo Single Tube Scanner for 2D barcodes and 1D linearbarcodes with Cryoprotection, Camera-based reader for 2Dbarcodes on the bottom of tubes with linear scanner for 1Drack codes
Catalog No. NC3585175 Supplier Azenta US Inc Supplier No. ZTSDUO80
Add to Cart
Add to Cart
Duo Single Tube Scanner for 2D barcodes and 1D linearbarcodes with Cryoprotection, Camera-based reader for 2Dbarcodes on the bottom of tubes with linear scanner for 1Drack codes