Learn More
Chemglass Life Sciences DuoCap, Sterile, 53-B, 48/cs
SDP
Catalog No. NC1586174
Encompass_Preferred
The DuoCap is designed for both aerobic and anaerobic applications. For aerobic applications the cap utilizes a 0.22um pore size PTFE membrane for sterile air exchange. For anaerobic applications the cap can easily be converted to a solid top design. Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. New EZ-Tear packaging allows for easy access. Caps are autoclavable.
This item is not returnable. View return policy
Catalog No. NC1586174 Supplier Chemglass Life Sciences Supplier No. CLS20900053M
The DuoCap is designed for both aerobic and anaerobic applications. For aerobic applications the cap utilizes a 0.22um pore size PTFE membrane for sterile air exchange. For anaerobic applications the cap can easily be converted to a solid top design. Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. New EZ-Tear packaging allows for easy access. Caps are autoclavable.