Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duocarmycin analog | 372954-15-9 | 95.9% | 611.09 g·mol⁻¹ | C34H31ClN4O5 | 1 MG
SDP

Catalog No. 5000203123
Encompass_Preferred

Duocarmycin analog is a synthetic analog of duocarmycin used as a DNA alkylator and as an antibody-drug conjugate (ADC) payload for research applications. Supplied as a solid powder, it is intended for laboratory research use only and not for human or clinical use.

  • Acts as a DNA alkylator and ADC cytotoxin.
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • Purity: 95.9%.
  • Cas number: 372954-15-9.
  • Molecular formula: C34H31ClN4O5; molecular weight: 611.09 g·mol⁻¹.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Appearance: off-white to light yellow solid.

Catalog No. 50-002-03123 Supplier Medchemexpress LLC Supplier No. HY1293551MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duocarmycin analog is a synthetic analog of duocarmycin used as a DNA alkylator and as an antibody-drug conjugate (ADC) payload for research applications. Supplied as a solid powder, it is intended for laboratory research use only and not for human or clinical use.

  • Acts as a DNA alkylator and ADC cytotoxin.
  • Available sizes: 1 mg, 5 mg, 10 mg.
  • Purity: 95.9%.
  • Cas number: 372954-15-9.
  • Molecular formula: C34H31ClN4O5; molecular weight: 611.09 g·mol⁻¹.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Appearance: off-white to light yellow solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.