Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duocarmycin DM free base | 1116745-06-2 | 463.96 g/mol | C26H26ClN3O3 | 5 MG
SDP

Catalog No. 5000202823
Encompass_Preferred

Duocarmycin DM free base is a DNA minor-groove alkylator used as a cytotoxic payload in antibody-drug conjugate (ADC) research and as a reagent for DNA-alkylation studies.

  • DNA minor-groove alkylator
  • Used as an ADC payload and research reagent for DNA alkylation studies
  • CAS number 1116745-06-2
  • Molecular formula C26H26ClN3O3
  • Molecular weight 463.96 g/mol
  • Appearance: light yellow to green yellow solid

Catalog No. 50-002-02823 Supplier Medchemexpress LLC Supplier No. HY1289155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duocarmycin DM free base is a DNA minor-groove alkylator used as a cytotoxic payload in antibody-drug conjugate (ADC) research and as a reagent for DNA-alkylation studies.

  • DNA minor-groove alkylator
  • Used as an ADC payload and research reagent for DNA alkylation studies
  • CAS number 1116745-06-2
  • Molecular formula C26H26ClN3O3
  • Molecular weight 463.96 g/mol
  • Appearance: light yellow to green yellow solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.