Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duocarmycin DM free base | 1116745-06-2 | MFCD32201125 | 99.7% | 463.96 | C26H26ClN3O3 | 10 MG
SDP

Catalog No. 5000203476
Encompass_Preferred

Duocarmycin DM free base is a DNA minor-groove alkylating cytotoxin used in antibody-drug conjugate (ADC) research and studies of DNA damage and cell-cycle effects. It alkylates adenine in AT-rich sequences and exhibits extremely potent cytotoxicity against cancer cells.

  • dna minor-groove alkylator used as an adc cytotoxin and research payload.
  • cas number 1116745-06-2; molecular formula C26H26ClN3O3; molecular weight 463.96.
  • high solubility in DMSO (50 mg/mL); in vivo formulations achieve ≥2 mg/mL as clear solutions.
  • storage: -20°C protected from light and under nitrogen; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • available pack sizes include 1 mg, 5 mg, and 10 mg for stock preparation.

Catalog No. 50-002-03476 Supplier Medchemexpress LLC Supplier No. HY12891510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duocarmycin DM free base is a DNA minor-groove alkylating cytotoxin used in antibody-drug conjugate (ADC) research and studies of DNA damage and cell-cycle effects. It alkylates adenine in AT-rich sequences and exhibits extremely potent cytotoxicity against cancer cells.

  • dna minor-groove alkylator used as an adc cytotoxin and research payload.
  • cas number 1116745-06-2; molecular formula C26H26ClN3O3; molecular weight 463.96.
  • high solubility in DMSO (50 mg/mL); in vivo formulations achieve ≥2 mg/mL as clear solutions.
  • storage: -20°C protected from light and under nitrogen; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • available pack sizes include 1 mg, 5 mg, and 10 mg for stock preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.