Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duocarmycin DM (free base) | 1116745-06-2 | MFCD32201125 | 99.7% | 463.96 | C26H26ClN3O3 | 50 MG
SDP

Catalog No. 5000203600
Encompass_Preferred

Duocarmycin DM (free base) is a potent DNA minor-groove alkylating agent used as a cytotoxic payload in antibody-drug conjugate research and DNA-damage studies. It is supplied as a solid with high purity and is soluble in DMSO for in vitro or formulation use.

  • Potent DNA minor-groove alkylator for ADC payload research.
  • Molecular weight 463.96 g/mol; formula C26H26ClN3O3.
  • High purity (99.7%) suitable for preclinical studies.
  • Soluble in DMSO (~50 mg/mL) and formulatable for in vivo use.
  • Store long term at -20°C; protect from light and oxygen.

Catalog No. 50-002-03600 Supplier Medchemexpress LLC Supplier No. HY12891550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duocarmycin DM (free base) is a potent DNA minor-groove alkylating agent used as a cytotoxic payload in antibody-drug conjugate research and DNA-damage studies. It is supplied as a solid with high purity and is soluble in DMSO for in vitro or formulation use.

  • Potent DNA minor-groove alkylator for ADC payload research.
  • Molecular weight 463.96 g/mol; formula C26H26ClN3O3.
  • High purity (99.7%) suitable for preclinical studies.
  • Soluble in DMSO (~50 mg/mL) and formulatable for in vivo use.
  • Store long term at -20°C; protect from light and oxygen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.